4-(2-Methyl-1H-benzo[d]imidazol-1-yl)benzoic acid ≥95%

Supplier: Matrix Scientific

057991-1G
101913-860EA 553.85 USD
101913-860
4-(2-Methyl-1H-benzo[d]imidazol-1-yl)benzoic acid ≥95%
4-(2-Methyl-1H-benzo[d]imidazol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₂N₂O₂
MDL Number: MFCD13810991
CAS Number: 1021144-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR