2,7-Dimethylimidazo[1,2-a]pyridine-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

057987-1G
101913-852EA 1147.7 USD
101913-852
2,7-Dimethylimidazo[1,2-a]pyridine-3-carbaldehyde ≥95%
2,7-Dimethylimidazo[1,2-a]pyridine-3-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MDL Number: MFCD03788999
CAS Number: 820245-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR