4-(4-(2-Bromoethoxy)phenyl)-4H-1,2,4-triazole ≥95%

Supplier: Matrix Scientific

057984-1G
101913-846EA 773.85 USD
101913-846
4-(4-(2-Bromoethoxy)phenyl)-4H-1,2,4-triazole ≥95%
4-(4-(2-Bromoethoxy)phenyl)-4H-1,2,4-triazole

Hazard: Irritant

Formula: C₁₀H₁₀BrN₃O
MDL Number: MFCD14708240
CAS Number: 1221279-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR