5-Methyl-3-(piperidin-3-yl)-1,2,4-oxadiazole ≥95%

Supplier: Matrix Scientific

057980-1G
101913-838EA 643.08 USD
101913-838
5-Methyl-3-(piperidin-3-yl)-1,2,4-oxadiazole ≥95%
5-Methyl-3-(piperidin-3-yl)-1,2,4-oxadiazole

Hazard: Irritant

Formula: C₈H₁₃N₃O
MDL Number: MFCD14707698
CAS Number: 895573-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR