N,N-Dimethyl-4-(pyrrolidin-3-yl)benzamide hydrochloride ≥95%

Supplier: Matrix Scientific

057977-1G
101913-834EA 2583.08 USD
101913-834
N,N-Dimethyl-4-(pyrrolidin-3-yl)benzamide hydrochloride ≥95%
N,N-Dimethyl-4-(pyrrolidin-3-yl)benzamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O
MDL Number: MFCD14708239
CAS Number: 1223748-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR