5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine ≥95%

Supplier: Matrix Scientific

057971-1G
101913-826EA 600 USD
101913-826
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine ≥95%
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine

Hazard: Irritant

Formula: C₅H₈N₄
MW: 124.144 g/mol
MDL Number: MFCD04115284
CAS Number: 837430-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR