6-Fluoro-2-methyl-2,3-dihydro-1H-inden-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Fluoro-2-methylindanone

057969-5G
101913-822EA 472.31 USD
101913-822
6-Fluoro-2-methyl-2,3-dihydro-1H-inden-1-one ≥95%
6-Fluoro-2-methyl-2,3-dihydro-1H-inden-1-one

Hazard: Irritant

Formula: C₁₀H₉FO
MW: 164.18 g/mol
MDL Number: MFCD00029129
CAS Number: 37794-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR