1H-Indazole-5-carboxylic acid ≥97%

Supplier: Matrix Scientific

057942-5G
101913-774EA 63.25 USD
101913-774
1H-Indazole-5-carboxylic acid ≥97%
1H-Indazole-5-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD06804570
CAS Number: 61700-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR