3,5-Di(1H-benzo[d]imidazol-2-yl)pyridine ≥98%

Supplier: Matrix Scientific

057932-5G
101913-754EA 953.85 USD
101913-754
3,5-Di(1H-benzo[d]imidazol-2-yl)pyridine ≥98%
3,5-Di(1H-benzo[d]imidazol-2-yl)pyridine

Hazard: Irritant

Formula: C₁₉H₁₃N₅
MDL Number: MFCD12545922
CAS Number: 111397-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR