2,6-Di(benzo[d]oxazol-2-yl)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Bis(benzo[d]oxazol-2-yl)pyridine

057931-5G
101913-750EA 1127.69 USD
101913-750
2,6-Di(benzo[d]oxazol-2-yl)pyridine ≥95%
2,6-Di(benzo[d]oxazol-2-yl)pyridine

Hazard: Irritant

Formula: C₁₉H₁₁N₃O₂
MW: 313.32 g/mol
MDL Number: MFCD05241117
CAS Number: 33858-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR