1,3-Di(benzo[d]oxazol-2-yl)benzene ≥95%

Supplier: Matrix Scientific

Synonyms: 1,3-Bis(benzo[d]oxazol-2-yl)benzene

057930-5G
101913-746EA 1115.38 USD
101913-746
1,3-Di(benzo[d]oxazol-2-yl)benzene ≥95%
1,3-Di(benzo[d]oxazol-2-yl)benzene

Hazard: Irritant

Formula: C₂₀H₁₂N₂O₂
MW: 312.33 g/mol
MDL Number: MFCD00382603
CAS Number: 59049-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR