5-Bromo-4-methylpyrimidine ≥95%

Supplier: Matrix Scientific

057914-1G
101913-730EA 167.69 USD
101913-730
5-Bromo-4-methylpyrimidine ≥95%
5-Bromo-4-methylpyrimidine

Hazard: Irritant

Formula: C₅H₅BrN₂
MW: 173.0100 g/mol
MDL Number: MFCD10000597
CAS Number: 1439-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR