2-Amino-5-pyrimidineboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine2-Aminopyrimidine-5-boronic acid Pinacol ester5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine2-(2-Aminopyrimidin-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

057913-5G
101913-726EA 67.69 USD
101913-726
2-Amino-5-pyrimidineboronic acid pinacol ester ≥95%
2-Amino-5-pyrimidineboronic acid pinacol ester

Hazard: Irritant-Harmful/Stored under Argon

Formula: C₁₀H₁₆BN₃O₂
MW: 221.06 g/mol
Melting Pt: 209…211 °C
MDL Number: MFCD06795685
CAS Number: 402960-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR