Ethyl-4-methylisoxazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-methylisoxazole-3-carboxylate

057885-250MG
102833-240EA 272.31 USD
102833-240
Ethyl-4-methylisoxazole-3-carboxylate ≥95%
Ethyl-4-methylisoxazole-3-carboxylate

Hazard: Irritant

Formula: C₇H₉NO₃
MW: 155.15 g/mol
MDL Number: MFCD14584426
CAS Number: 38061-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR