5-(1H-Pyrrol-2-yl)-1H-tetrazole ≥98%

Supplier: Matrix Scientific

057870-1G
101913-676EA 281.54 USD
101913-676
5-(1H-Pyrrol-2-yl)-1H-tetrazole ≥98%
5-(1H-Pyrrol-2-yl)-1H-tetrazole

Hazard: Irritant

Formula: C₅H₅N₅
MDL Number: MFCD12828257
CAS Number: 31602-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR