3-(7-Cyano-1H-indol-3-yl)propanoic acid ≥95%

Supplier: Matrix Scientific

057851-1G
101913-672EA 1187.69 USD
101913-672
3-(7-Cyano-1H-indol-3-yl)propanoic acid ≥95%
3-(7-Cyano-1H-indol-3-yl)propanoic acid

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₂
MDL Number: MFCD14708232
CAS Number: 1223748-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR