1H-Indole-7-carbonitrile ≥95%

Supplier: Matrix Scientific

057849-1G
101913-668EA 60 USD
101913-668
1H-Indole-7-carbonitrile ≥95%
1H-Indole-7-carbonitrile

Hazard: Irritant

Formula: C₉H₆N₂
MW: 142.1573 g/mol
MDL Number: MFCD00800653
CAS Number: 96631-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR