4,7-Dibromo-1H-indole ≥95%

Supplier: Matrix Scientific

057848-1G
101913-664EA 1126.15 USD
101913-664
4,7-Dibromo-1H-indole ≥95%
4,7-Dibromo-1H-indole

Hazard: Irritant

Formula: C₈H₅Br₂N
MDL Number: MFCD08272255
CAS Number: 126811-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR