1-Methyl-3-(1-methylpiperidin-4-yl)-1H-indol-5-ol ≥95%

Supplier: Matrix Scientific

057846-1G
101913-660EA 946.15 USD
101913-660
1-Methyl-3-(1-methylpiperidin-4-yl)-1H-indol-5-ol ≥95%
1-Methyl-3-(1-methylpiperidin-4-yl)-1H-indol-5-ol

Hazard: Irritant

Formula: C₁₅H₂₀N₂O
MDL Number: MFCD14708231
CAS Number: 445441-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR