Ethyl-5-amino-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-Aminoindole-2-carboxylate

057844-5G
101913-656EA 784.62 USD
101913-656
Ethyl-5-amino-1H-indole-2-carboxylate ≥95%
Ethyl-5-amino-1H-indole-2-carboxylate

Hazard: Irritant/Stored under Argon

Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
MDL Number: MFCD08689973
CAS Number: 71086-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR