(S)-Benzyl-3-hydroxy-1-(methylamino)-1-oxopropan-2-ylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-Benzyl (3-hydroxy-1-(methylamino)-1-oxopropan-2-yl)carbamate

057766-5G
101913-604EA 753.85 USD
101913-604
(S)-Benzyl-3-hydroxy-1-(methylamino)-1-oxopropan-2-ylcarbamate ≥95%
(S)-Benzyl-3-hydroxy-1-(methylamino)-1-oxopropan-2-ylcarbamate

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₄
MW: 252.27 g/mol
MDL Number: MFCD17011849
CAS Number: 19647-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR