(R)-tert-Butyl-1-(methoxy(methyl)amino)-1-oxopropan-2-ylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-tert-Butyl (1-(methoxy(methyl)amino)-1-oxopropan-2-yl)carbamate

057765-5G
102551-496EA 287.69 USD
102551-496
(R)-tert-Butyl-1-(methoxy(methyl)amino)-1-oxopropan-2-ylcarbamate ≥95%
(R)-tert-Butyl-1-(methoxy(methyl)amino)-1-oxopropan-2-ylcarbamate

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₄
MW: 232.28 g/mol
MDL Number: MFCD10698129
CAS Number: 146553-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR