Chlorphenoxamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1-(4-Chlorophenyl)-1-phenylethoxy)-N,N-dimethylethan-1-amine

057751-1G
101913-588EA 361.54 USD
101913-588
Chlorphenoxamine ≥95%
Chlorphenoxamine

Hazard: Irritant

Formula: C₁₈H₂₂ClNO
MW: 303.84 g/mol
MDL Number: MFCD00866588
CAS Number: 77-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR