3-(Dimethylamino)-4-methoxybenzoic acid ≥95%

Supplier: Matrix Scientific

057749-1G
101913-584EA 735.38 USD
101913-584
3-(Dimethylamino)-4-methoxybenzoic acid ≥95%
3-(Dimethylamino)-4-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₀H₁₃NO₃
MDL Number: MFCD16659617
CAS Number: 197500-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR