2-Bromo-p-anisidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4-methoxyaniline

057748-1G
101913-580EA 16.1 USD
101913-580
2-Bromo-p-anisidine ≥95%
2-Bromo-p-anisidine

Hazard: Toxic/Store in Freezer

Formula: C₇H₈BrNO
MW: 202.0485 g/mol
MDL Number: MFCD09029057
CAS Number: 32338-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR