4-Bromo-3,5-dichlorofluorobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1,3-dichloro-5-fluorobenzene

057714-25G
101913-570EA 638.46 USD
101913-570
4-Bromo-3,5-dichlorofluorobenzene ≥95%
4-Bromo-3,5-dichlorofluorobenzene

Hazard: Irritant

Formula: C₆H₂BrCl₂F
MW: 243.9 g/mol
MDL Number: MFCD11845890
CAS Number: 263333-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR