Methyl-3-(cyanomethyl)benzoate ≥95%

Synonyms: Methyl 3-(cyanomethyl)benzoate

057695-25G
101913-548EA 1120.63 USD
101913-548
Methyl-3-(cyanomethyl)benzoate ≥95%
Methyl-3-(cyanomethyl)benzoate

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
MDL Number: MFCD07783720
CAS Number: 68432-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR