2-(2-Bromophenyl)-2-oxoacetic acid ≥95%

Supplier: Matrix Scientific

057694-5G
101913-544EA 738.46 USD
101913-544
2-(2-Bromophenyl)-2-oxoacetic acid ≥95%
2-(2-Bromophenyl)-2-oxoacetic acid

Hazard: Irritant

Formula: C₈H₅BrO₃
MDL Number: MFCD00463413
CAS Number: 26767-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR