3-(Hydroxymethyl)benzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Cyanobenzyl alcohol

057673-1G
101913-522EA 158.46 USD
101913-522
3-(Hydroxymethyl)benzonitrile ≥95%
3-(Hydroxymethyl)benzonitrile

Hazard: Irritant

Formula: C₈H₇NO
MW: 133.15 g/mol
MDL Number: MFCD01321087
CAS Number: 874-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR