(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodophenylamino)benzamide ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-((2-fluoro-4-iodophenyl)amino)benzamidePD-0325901

057665-100MG
101913-514EA 793.85 USD
101913-514
(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodophenylamino)benzamide ≥95%
(R)-N-(2,3-Dihydroxypropoxy)-3,4-difluoro-2-(2-fluoro-4-iodophenylamino)benzamide

Hazard: Irritant

Formula: C₁₆H₁₄F₃IN₂O₄
MW: 482.2 g/mol
MDL Number: MFCD08435926
CAS Number: 391210-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR