Methyl-4-(2-bromoethyl)benzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(2-bromoethyl)benzoate

057646-1G
101913-472EA 230.77 USD
101913-472
Methyl-4-(2-bromoethyl)benzoate ≥95%
Methyl-4-(2-bromoethyl)benzoate

Hazard: Irritant

Formula: C₁₀H₁₁BrO₂
MW: 243.11 g/mol
MDL Number: MFCD14708225
CAS Number: 136333-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR