2-(4-Fluorophenyl)propan-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: alpha, alpha-Dimethyl-4-fluorobenzylamine1-(4-Fluorophenyl)-1-methylethylamine

057639-1G
101913-462EA 344.62 USD
101913-462
2-(4-Fluorophenyl)propan-2-amine ≥95%
2-(4-Fluorophenyl)propan-2-amine

Hazard: Corrosive/Stored under Argon/Air Sensitive/Refrigerate

Formula: C₉H₁₂FN
MW: 153.2 g/mol
MDL Number: MFCD07777176
CAS Number: 17797-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR