Methyl-4-iodo-3-methoxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-iodo-3-methoxybenzoate

057634-1G
101913-450EA 83.08 USD
101913-450
Methyl-4-iodo-3-methoxybenzoate ≥95%
Methyl 4-iodo-3-methoxybenzoate

Hazard: Irritant

Formula: C₉H₉IO₃
MW: 292.0704 g/mol
MDL Number: MFCD06203960
CAS Number: 35387-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR