2-Chloro-6-formylbenzonitrile ≥95%

Supplier: Matrix Scientific

057632-1G
101913-446EA 415.38 USD
101913-446
2-Chloro-6-formylbenzonitrile ≥95%
2-Chloro-6-formylbenzonitrile

Hazard: Irritant

Formula: C₈H₄ClNO
MDL Number: MFCD10699534
CAS Number: 77532-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR