3-Anilinopropionitrile ≥95%

Supplier: Matrix Scientific

057627-1G
101913-438EA 287.5 USD
101913-438
3-Anilinopropionitrile ≥95%
3-Anilinopropionitrile

Hazard: Irritant

Formula: C₉H₁₀N₂
MDL Number: MFCD00001953
CAS Number: 1075-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR