(2S,3R,4S,5S)-2-(4-Nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol ≥95%

Supplier: Matrix Scientific

Synonyms: p-Nitrophenyl alpha-L-arabinopyranoside

057624-1G
101913-434EA 1306.15 USD
101913-434
(2S,3R,4S,5S)-2-(4-Nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol ≥95%
(2S,3R,4S,5S)-2-(4-Nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

Hazard: Irritant

Formula: C₁₁H₁₃NO₇
MW: 271.23 g/mol
MDL Number: MFCD00151486
CAS Number: 1223-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR