3,5-Di-tert-butylphenyl acrylate ≥95%

Supplier: Matrix Scientific

057617-1G
101913-420EA 1658.46 USD
101913-420
3,5-Di-tert-butylphenyl acrylate ≥95%
3,5-Di-tert-butylphenyl acrylate

Hazard: Irritant

Formula: C₁₇H₂₄O₂
MDL Number: MFCD15072119
CAS Number: 1223748-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR