5-Cyano-2-fluorobenzaldehyde ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Fluoro-3-formylbenzonitrile

057614-500MG
101913-412EA 32.31 USD
101913-412
5-Cyano-2-fluorobenzaldehyde ≥98%
5-Cyano-2-fluorobenzaldehyde

Hazard: Irritant

Formula: C₈H₄FNO
MW: 149.1219 g/mol
MDL Number: MFCD04974126
CAS Number: 146137-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR