Methyl-3-amino-4-bromobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-4-bromobenzoate

057605-5G
101913-392EA 36.92 USD
101913-392
Methyl-3-amino-4-bromobenzoate ≥95%
Methyl 3-amino-4-bromobenzoate

Hazard: Irritant

Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
MDL Number: MFCD00553090
CAS Number: 46064-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR