(9H-Fluoren-9-yl)methyl-4-aminophenylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: (9H-Fluoren-9-yl)methyl (4-aminophenyl)carbamate

057603-5G
101913-388EA 961.54 USD
101913-388
(9H-Fluoren-9-yl)methyl-4-aminophenylcarbamate ≥95%
(9H-Fluoren-9-yl)methyl-4-aminophenylcarbamate

Hazard: Irritant

Formula: C₂₁H₁₈N₂O₂
MW: 330.39 g/mol
MDL Number: MFCD12912141
CAS Number: 205688-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR