1-Benzyl-1,2,3,4-tetrahydroisoquinoline ≥96%

Supplier: Matrix Scientific

057594-1G
101913-364EA 1230.77 USD
101913-364
1-Benzyl-1,2,3,4-tetrahydroisoquinoline ≥96%
1-Benzyl-1,2,3,4-tetrahydroisoquinoline

Hazard: Irritant

Formula: C₁₆H₁₇N
MDL Number: MFCD00024130
CAS Number: 19716-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR