3-Chloro-3-cyclopentylpropionyl chloride ≥95%

Supplier: Matrix Scientific

057591-1G
101913-356EA 1401.54 USD
101913-356
3-Chloro-3-cyclopentylpropionyl chloride ≥95%
3-Chloro-3-cyclopentylpropionyl chloride

Hazard: Irritant

Formula: C₈H₁₂Cl₂O
MDL Number: MFCD16877033
CAS Number: 1253790-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR