(1R,4R)-4-(1,1-Dimethylethylsulfonamido)cyclohexanecarboxylic acid ≥95%

Supplier: Matrix Scientific

057576-1G
101913-336EA 2147.69 USD
101913-336
(1R,4R)-4-(1,1-Dimethylethylsulfonamido)cyclohexanecarboxylic acid ≥95%
(1R,4R)-4-(1,1-Dimethylethylsulfonamido)cyclohexanecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₂₁NO₄S
MDL Number: MFCD17011838
CAS Number: 342578-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR