Monomethyl-trans-1,4-cyclohexanedicarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
trans-1,4-Cyclohexanedicarboxylic Acid Monomethyl Ester
, trans-4-(Methoxycarbonyl)cyclohexanecarboxylic acid
, Methyl Hydrogen trans-1,4-Cyclohexanedicarboxylate
057573-5G
101913-332EA
29.23
USD
101913-332
Monomethyl-trans-1,4-cyclohexanedicarboxylate ≥95%
Monomethyl-trans-1,4-cyclohexanedicarboxylate
Purity: 95%
CAS: 15177-67-0
MDL Number: MFCD01311247
Molecular Formula: C9H14O4
Molecular Weight: 186,21
Hazard: Irritant
Formula:
C₉H₁₄O₄ MW: 186.21 g/mol Melting Pt: 124…128 °C |
MDL Number:
MFCD01311247 CAS Number: 15177-67-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...