Methyl-2-amino-2-cyclopropylacetate hydrochloride ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-2-cyclopropylacetate hydrochloride

057568-1G
101913-322EA 183.08 USD
101913-322
Methyl-2-amino-2-cyclopropylacetate hydrochloride ≥98%
Methyl-2-amino-2-cyclopropylacetate hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO₂
MW: 165.62 g/mol
MDL Number: MFCD12031219
CAS Number: 535936-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR