2-(Chloromethyl)-1-methyl-1H-benzo[d]imidazole ≥95%
Supplier: Matrix Scientific
057565-500MG
101913-314EA
184.62
USD
101913-314
2-(Chloromethyl)-1-methyl-1H-benzo[d]imidazole ≥95%
2-(Chloromethyl)-1-methyl-1H-benzo[d]imidazole
Hazard: Irritant
Formula:
C₉H₉ClN₂ MW: 180.64 g/mol |
MDL Number:
MFCD00457523 CAS Number: 4760-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...