1-Methylimidazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methylimidazole-5-carboxylic Acid

057544-1G
101913-276EA 15.38 USD
101913-276
1-Methylimidazole-5-carboxylic acid ≥95%
1-Methylimidazole-5-carboxylic acid

Hazard: Irritant

Formula: C₅H₆N₂O₂
MW: 126.1133 g/mol
MDL Number: MFCD00955677
CAS Number: 41806-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR