1-Methylimidazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Methylimidazole-5-carboxaldehyde1-Methyl-1H-imidazole-5-carbaldehyde

057543-1G
101913-272EA 152.31 USD
101913-272
1-Methylimidazole-5-carbaldehyde ≥95%
1-Methylimidazole-5-carbaldehyde

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD00030439
CAS Number: 39021-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR