Methyl methyl-4-nitro-1H-imidazole-2-carboxylate ≥98%

Supplier: Matrix Scientific

Synonyms: Methyl 1-methyl-4-nitro-1H-imidazole-2-carboxylate

057542-1G
101913-268EA 218.46 USD
101913-268
Methyl methyl-4-nitro-1H-imidazole-2-carboxylate ≥98%
Methyl methyl-4-nitro-1H-imidazole-2-carboxylate

Hazard: Irritant

Formula: C₆H₇N₃O₄
MW: 185.14 g/mol
MDL Number: MFCD08275078
CAS Number: 169770-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR