5-Chloro-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

057534-1G
101913-248EA 769.23 USD
101913-248
5-Chloro-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid ≥95%
5-Chloro-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrrolo[2,3-c]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₉ClN₂O₃Si
MDL Number: MFCD14708217
CAS Number: 1223748-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR